Synonym: cis,cis-11,14-Eicosadienoic acid diepoxide methyl ester
Empirical Formula (Hill Notation) C21H38O4
Molecular Weight 354.52
MDL number MFCD07370104
PubChem Substance ID 24866896
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Biochemicals and Reagents, Chemical Class, |
| grade | qualitative standard |
| assay | ≥97.0% (sum of stereoisomers, GC) |
| storage temp. | −20°C |
Internal standard for the assay of epoxidized soy bean oil (ESBO) in foods with HPLC-MS or GC-MS. ESBO is currently still employed as plasticizer and HCl scavenger in foamed PVC used in cap linings
Customers Also Viewed
analytical standard, for food analysis
analytical standard, for food analysis
dietary source of long-chain triglycerides and other lipids
contains 4,000 ppm monomethyl ether hydroquinone as inhibitor
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?