EMAIL THIS PAGE TO A FRIEND

43728 Fluka

BTEX Solution

analytical standard

Purchase

Properties

Related Categories A-C, Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments,
grade   analytical standard
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.480
density   1.099 g/mL at 20 °C
format   multi-component solution
storage temp.   2-8°C

Description

Analysis Note

solution contains benzene, toluene, ethylbenzene, p-xylene, m-xylene, o-xylene, 1 mg/ml each in DMSO

Price and Availability


Explore the New Analytical Standards Portal

Customers Also Viewed

10 ppm each component in nitrogen, analytical standard

certified reference material, 2000 μg/mL each component in methanol, ampule of 1 mL

certified reference material, TraceCERT®, 2000 μg/mL each component in methanol

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?