Synonym: 2,2′-Dipyridyl disulfide
CAS Number 2127-03-9
Empirical Formula (Hill Notation) C10H8N2S2
Molecular Weight 220.31
Beilstein Registry Number 154629
EC Number 218-343-1
MDL number MFCD00006287
Popular Documents: FTNMR (PDF)
| grade | purum |
| assay | ≥97.0% (GC) |
| mp | 56-58 °C(lit.) |
| 56-58 °C | |
| storage temp. | 2-8°C |
Reagent used for the lactonization of hydroxy acids with PPh31; Reagent used for sulfenylations
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?