43942 Fluka

Docosane

analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 629-97-0

  • Linear Formula CH3(CH2)20CH3

  • Molecular Weight 310.60

  •  Beilstein Registry Number 1702206

  •  EC Number 211-121-5

  •  MDL number MFCD00009348

  •  PubChem Substance ID 24867506

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, D,
grade   analytical standard
vapor density   10.8 (vs air)
vapor pressure   <1 mmHg ( 21.1 °C)
assay   ≥99.5% (GC)
shelf life   (limited shelf life, expiry date on the label)
bp   369 °C(lit.)
mp   42-45 °C(lit.)
  43-46 °C
density   0.778 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Docosane
134457Docosane

99%

Docosane
442670Docosane

analytical standard

Eicosane

analytical standard

Heneicosane

analytical standard

Tetracosane

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?