Synonym: Behenyl alcohol
CAS Number 661-19-8
Linear Formula CH3(CH2)21OH
Molecular Weight 326.60
Beilstein Registry Number 1770470
EC Number 211-546-6
MDL number MFCD00002939
PubChem Substance ID 24867521
Popular Documents: FTNMR (PDF)
| Related Categories | Alcohols, Biochemicals and Reagents, Building Blocks, C11 to C30+, Chemical Synthesis, |
| grade | purum |
| assay | ≥97.0% (GC) |
| bp | 180 °C/0.22 mmHg(lit.) |
| mp | 65-72 °C(lit.) |
| 70-71 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?