Synonym:
3,3′,4′,5,7-Pentahydroxyflavone 3-O-α-
CAS Number 572-30-5
Empirical Formula (Hill Notation) C20H18O11
Molecular Weight 434.35
MDL number MFCD10566630
| Related Categories | A - D, Alphabetical, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | analytical standard |
| assay | ≥85% (HPLC) |
| shelf life | (limited shelf life, expiry date on the label) |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?