Synonym: Lauryltrimethylammonium bromide
CAS Number 1119-94-4
Linear Formula CH3(CH2)11N(CH3)3Br
Molecular Weight 308.34
Beilstein Registry Number 3597463
EC Number 214-290-3
MDL number MFCD00011767
PubChem Substance ID 24867824
Popular Documents: RAMAN FTIR (PDF)
| Related Categories | Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Cationic Detergents, |
| grade | for ion pair chromatography |
| description | cationic |
| assay | ≥99.0% (AT) |
| mp | 246 °C (dec.)(lit.) |
| λ | 10 % in H2O |
| UV absorption | λ: 240 nm Amax: ≤0.2 |
| λ: 250 nm Amax: ≤0.03 |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?