• USA Home
  • 443093 - Filling solution for sodium electrode (2M NH4Cl)

EMAIL THIS PAGE TO A FRIEND

443093 Fluka

Filling solution for sodium electrode (2M NH4Cl)

Purchase

Properties

bp   100 °C/750 mmHg
density   1.03 g/mL at 25 °C

Description

General description

Visit our Sensor Applications portal to learn more.

Price and Availability

Customers Also Viewed

Ammonium chloride

99.998% trace metals basis

Safety & Documentation

Safety Information

Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?