EMAIL THIS PAGE TO A FRIEND

44578 Fluka

Dragendorff reagent

for TLC derivatization

Purchase

Description

Application

spray and plunge reagent for chromatography

Other Notes

This non-aqueous Thiess, Reuther and Vagujfalvi variant of the reagent is used to detect alkaloids and other nitrogen compounds in TLC1,2,3

Price and Availability


Explore the New Analytical Standards Portal

Additional Products to Explore

Bismuth(III) nitrate pentahydrate

ACS reagent, ≥98.0%

Methyl 4-hydroxybenzoate

ReagentPlus®, ≥99.0%, crystalline

Vanillin

ReagentPlus®, 99%

Customers Also Viewed

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
44.6 °F
Flash Point(C) 
7 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. A. Baerheim Svendsen J. Planar Chromatogr. 2, 8, (1989)

2. W. Funk, S. Dröschel J. Planar Chromatogr. 4, 123, (1991)

3. E. Talas et al. J. Planar Chromatogr. 5, 28, (1992)

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?