EMAIL THIS PAGE TO A FRIEND

44660 Fluka

Fast Blue B Salt

for microscopy (Hist.), for TLC-detection of aflatoxins

Synonym: o-Dianisidine bis(diazotized) zinc double salt, Azoic Diazo No. 48, DBB, Diazo Blue B, Naphthanil Diazo Blue B

  • CAS Number 14263-94-6

  • Linear Formula C14H12N4O2Cl2 · ZnCl2

  • Molecular Weight 475.47

  •  Beilstein Registry Number 1499067

  •  Colour Index Number 37235

  •  EC Number 238-153-2

  •  MDL number MFCD00075203

  • Popular Documents:  RAMAN FTIR (PDF)  

Purchase

Properties

grade   for microscopy (Hist.)
quality   for TLC-detection of aflatoxins
mp   >300 °C(lit.)

Description

Other Notes

Stain used for the determination of various compounds, e.g. proteins1; bilirubin2; enzymes3;

This product has been replaced by D9805-Sigma-Aldrich | Fast Blue B Salt Dye content ~95 %

Price and Availability

Customers Also Viewed

Safety & Documentation

Safety Information

Hazard Codes (Europe) 
T
Risk Statements (Europe) 
45
Safety Statements (Europe) 
53.1-45
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Products

replaced by

Product #

Description

Add to Cart

D9805 Fast Blue B Salt, Dye content ~95 %

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?