CAS Number 112-95-8
Linear Formula CH3(CH2)18CH3
Molecular Weight 282.55
Beilstein Registry Number 1700722
EC Number 204-018-1
MDL number MFCD00009344
PubChem Substance ID 24868388
Popular Documents: FTNMR (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, E, |
| grade | analytical standard |
| vapor density | 9.8 (vs air) |
| assay | ≥99.5% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| bp | 220 °C/30 mmHg(lit.) |
| mp | 35-37 °C(lit.) |
| 37-40 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?