Synonym: Arachinyl alcohol
CAS Number 629-96-9
Linear Formula CH3(CH2)19OH
Molecular Weight 298.55
Beilstein Registry Number 1705104
EC Number 211-119-4
MDL number MFCD00002938
PubChem Substance ID 24868418
Popular Documents: FTNMR (PDF)
| Related Categories | Alcohols, Biochemicals and Reagents, Building Blocks, C11 to C30+, Chemical Synthesis, |
| vapor density | 7.4 (vs air) |
| vapor pressure | <0.1 mmHg ( 20 °C) |
| grade | purum |
| assay | ≥97.0% (GC) |
| mp | 62-65 °C(lit.) |
| 63-66 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?