Synonym: Ferrous gluconate hydrate
CAS Number 699014-53-4
Linear Formula (C6H11O7)2Fe · aq
Molecular Weight 446.14 (anhydrous basis)
Beilstein Registry Number 3805721
MDL number MFCD04037236
PubChem Substance ID 24868482
| Related Categories | Essential Chemicals, Inorganic Salts, Solutions and Reagents, Reagent Grade, Research Essentials |
| grade | purum p.a. |
| loss | 5-15% loss on drying |
| anion traces | chloride (Cl-): ≤500 mg/kg |
| sulfate (SO42-): ≤500 mg/kg |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?