Synonym: Elaidic acid methyl ester, Methyl trans-9-octadecenoate
CAS Number 1937-62-8
Linear Formula CH3(CH2)7CH=CH(CH2)7COOCH3
Molecular Weight 296.49
Beilstein Registry Number 1727038
EC Number 217-712-4
MDL number MFCD00066521
PubChem Substance ID 24868609
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Biochemicals and Reagents, Chromatography, |
| grade | analytical standard |
| assay | ≥99.0% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| refractive index |
n |
| mp | 9-10 °C |
| density | 0.871 g/mL at 20 °C(lit.) |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?