Synonym:
2-
CAS Number 834-12-8
Empirical Formula (Hill Notation) C9H17N5S
Molecular Weight 227.33
Beilstein Registry Number 613099
EC Number 212-634-7
MDL number MFCD00055398
PubChem Substance ID 24868770
| Related Categories | A, A-B, AM to AQ, All alphabetically sorted, Alphabetical Index of Analytical Standards, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?