Synonym: Guthion®, Methyltriazotin
CAS Number 86-50-0
Empirical Formula (Hill Notation) C10H12N3O3PS2
Molecular Weight 317.32
Beilstein Registry Number 280476
EC Number 201-676-1
MDL number MFCD00041814
PubChem Substance ID 24868791
| Related Categories | A, A-B, AR to AZ, Acaricides, All alphabetically sorted, |
| grade | PESTANAL®, analytical standard |
| storage temp. | 2-8°C |
Guthion is a registered trademark of Makhteshim Chemical Works Ltd.
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. 45333.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?