Synonym: Chlorpyriphos
CAS Number 2921-88-2
Empirical Formula (Hill Notation) C9H11Cl3NO3PS
Molecular Weight 350.59
Beilstein Registry Number 1545756
EC Number 220-864-4
MDL number MFCD00041800
PubChem Substance ID 24868870
| Related Categories | Acaricides, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
| storage temp. | 2-8°C |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?