Synonym:
(±)-3-(α-Acetonyl-p-
CAS Number 81-82-3
Empirical Formula (Hill Notation) C19H15ClO4
Molecular Weight 342.77
Beilstein Registry Number 6819431
EC Number 201-378-1
MDL number MFCD00012094
PubChem Substance ID 24868875
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, C, |
| grade | PESTANAL®, analytical standard |
| mp | 168-170 °C(lit.) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?