Synonym:
Succinic acid mono(2,2-
CAS Number 1596-84-5
Linear Formula HO2CCH2CH2CONHN(CH3)2
Molecular Weight 160.17
Beilstein Registry Number 1863230
EC Number 216-485-9
MDL number MFCD00002787
PubChem Substance ID 24868896
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
| mp | 162-164 °C(lit.) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?