CAS Number 52918-63-5
Empirical Formula (Hill Notation) C22H19Br2NO3
Molecular Weight 505.20
Beilstein Registry Number 6746312
EC Number 258-256-6
MDL number MFCD00870122
PubChem Substance ID 24868901
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?