Synonym:
3-
CAS Number 330-54-1
Empirical Formula (Hill Notation) C9H10Cl2N2O
Molecular Weight 233.09
Beilstein Registry Number 2215168
EC Number 206-354-4
MDL number MFCD00018136
PubChem Substance ID 24868945
| Related Categories | 2000/60/EC, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
| Gene Information |
human ... EPHX2(2053) mouse ... Ephx2(13850) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?