CAS Number 30043-49-3
Empirical Formula (Hill Notation) C7H12N4O3S2
Molecular Weight 264.33
Beilstein Registry Number 551847
EC Number 250-010-6
MDL number MFCD00078690
PubChem Substance ID 24868962
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?