Synonym: 2-Hydroxybiphenyl
CAS Number 90-43-7
Linear Formula C6H5C6H4OH
Molecular Weight 170.21
Beilstein Registry Number 606907
E Number E232
EC Number 201-993-5
MDL number MFCD00002208
PubChem Substance ID 24869050
| Related Categories | 2000/60/EC, All Alphabetically Sorted, All alphabetically sorted, Alphabetical, Alphabetical Index of Analytical Standards, |
| grade | PESTANAL®, analytical standard |
| vapor pressure | 7 mmHg ( 140 °C) |
| bp | 282 °C(lit.) |
| mp | 57-59 °C(lit.) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?