Synonym:
2-
CAS Number 96-45-7
Empirical Formula (Hill Notation) C3H6N2S
Molecular Weight 102.16
Beilstein Registry Number 106275
EC Number 202-506-9
MDL number MFCD00005276
PubChem Substance ID 24869052
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
| mp | 196-200 °C(lit.) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?