Synonym: Mordant Black 3
CAS Number 3564-14-5
Empirical Formula (Hill Notation) C20H13N2NaO5S
Molecular Weight 416.38
Colour Index Number 14640
EC Number 222-639-6
MDL number MFCD00003934
PubChem Substance ID 24869065
Popular Documents: FTIR (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Reagents, Analytical Standards, Analytical/Chromatography, E, |
| grade | indicator (for complexometry) |
Visit our Titration Center to learn more.
25 g in poly bottle
Eriochrome is a registered trademark of Huntsman International LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?