Synonym:
1-
CAS Number 2538-85-4
Empirical Formula (Hill Notation) C20H13N2NaO5S
Molecular Weight 416.38
Beilstein Registry Number 6041435
Colour Index Number 15705
EC Number 219-810-2
MDL number MFCD00004069
PubChem Substance ID 24869074
Popular Documents: FTIR (PDF)
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Reagents, Analytical Standards, Analytical/Chromatography, C, |
| grade | indicator (for complexometry (Al, Fe, Zr)) |
Visit our Titration Center to learn more.
Calcon is a trademark of Benckiser
Eriochrome is a registered trademark of Huntsman International LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?