• USA Home
  • 456098 - Buffer solution pH 2.0 (20°C)

EMAIL THIS PAGE TO A FRIEND
456098 Fluka

Buffer solution pH 2.0 (20°C)

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Buffer Solutions, Buffer Solutions, pH 1 - 4, Titration,
pH   2.00±0.02
bp   98.5-99.5 °C/740 mmHg(lit.)

Description

General description

Visit our Titration Center to learn more.

Packaging

500 mL in poly bottle

Preparation Note

Buffer preparation based on 3.7 g/L Potassium chloride and 0.95 g/L Hydrogen chloride. Final concentrations can vary slightly, as pH is adjusted after preparation.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
nwg
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?