CAS Number 24579-73-5
Empirical Formula (Hill Notation) C9H20N2O2
Molecular Weight 188.27
Beilstein Registry Number 2080745
MDL number MFCD00078717
PubChem Substance ID 24869170
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Carbamates, |
| grade | PESTANAL®, analytical standard |
| storage temp. | 2-8°C |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?