EMAIL THIS PAGE TO A FRIEND

45640 Fluka

Propazine

PESTANAL®, analytical standard

Synonym: 2,4-Bis(isopropylamino)-6-chloro-1,3,5-triazine

  • CAS Number 139-40-2

  • Empirical Formula (Hill Notation) C9H16ClN5

  • Molecular Weight 229.71

  •  Beilstein Registry Number 747081

  •  EC Number 205-359-9

  •  MDL number MFCD00047341

  •  PubChem Substance ID 24869173

Purchase

Description

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

QuEChERS SPE Tubes


QuEChERS SPE TubesSupelco provides a range of routine and specialty products for this popular sample prep technique used in multi-pesticide residue analysis.




Solvents for Pesticide Residue Analysis


Solvents for Pesticide Residue AnalysisFluka solvents for residue analysis of pesticides are glass-distilled and
extensively tested by GC/ECD to ensure minimum halogenated residue.




Customers Also Viewed

Prometon

PESTANAL®, analytical standard

Prometryn

PESTANAL®, analytical standard

Propazine

analytical standard, ampule of 100 mg

Propazine solution

100 ng/μL in methanol, PESTANAL®, analytical standard

Simazine

PESTANAL®, analytical standard

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
36/37-60-61
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
XY5300000

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Effects of chloro-s-triazine herbicides and metabolites on aromatase activity in various human cell lines and on vitellogenin production in male carp hepatocytes. Sanderson JT, Letcher RJ, Heneweer M, et al. Environ. Health Perspect. 109(10), 1027-31, (2001)

Loading...


Alteration of catecholamines in pheochromocytoma (PC12) cells in vitro by the metabolites of chlorotriazine herbicide. Das PC, McElroy WK, and Cooper RL Toxicol. Sci. 59(1), 127-37, (2001)

Loading...


"Does atrazine transform to propazine in aquatic sediments?": Comment on "biological and chemical transformations of atrazine in coastal aquatic sediments" by Kelly L. Smalling and C. Marjorie Aelion [Chemosphere 62 (2006) 188-196]. Thurman EM Chemosphere 65(8), 1440-1; discussion 1442-3, (2006)

Loading...


Molecular recognition in a propazine-imprinted polymer and its application to the determination of triazines in environmental samples. Turiel E, Martin-Esteban A, Fernández P, et al. Anal. Chem. 73(21), 5133-41, (2001)

Loading...


Determination of pesticides in surface and ground waters by liquid chromatography-electrospray-tandem mass spectrometry. Dujaković N, Grujić S, Radisić M, et al. Anal. Chim. Acta 678(1), 63-72, (2010)

Loading...


Occurrence and removal of chloro-s-triazines in water treatment plants. Jiang H, Adams C, Graziano N, et al. Environ. Sci. Technol. 40(11), 3609-16, (2006)

Loading...


Comparison of a non-aqueous capillary electrophoresis method with high performance liquid chromatography for the determination of herbicides and metabolites in water samples. Carabias-Martínez R, Rodríguez-Gonzalo E, Miranda-Cruz E, et al. J. Chromatogr. A 1122(1-2), 194-201, (2006)

Loading...


Computational design and synthesis of a high selective molecularly imprinted polymer for voltammetric sensing of propazine in food samples. Gholivand MB, Karimian N, and Malekzadeh G Talanta 89, 513-20, (2012)

Loading...


Determination of triazines and dealkylated and hydroxylated metabolites in river water using a propazine-imprinted polymer. Carabias-Martínez R, Rodríguez-Gonzalo E, and Herrero-Hernández E J. Chromatogr. A 1085(2), 199-206, (2005)

Loading...


Pubertal development in female Wistar rats following exposure to propazine and atrazine biotransformation by-products, diamino-S-chlorotriazine and hydroxyatrazine. Laws SC, Ferrell JM, Stoker TE, et al. Toxicol. Sci. 76(1), 190-200, (2003)

Loading...


A chemometrics approach for simultaneous determination of cyanazine and propazine based on a carbon paste electrode modified by a molecularly imprinted polymer. Gholivand MB, Shariati-Rad M, Karimian N, et al. Analyst 137(5), 1190-8, (2012)

Loading...


Characterisation and quality assessment of binding sites on a propazine-imprinted polymer prepared by precipitation polymerisation. Cacho C, Turiel E, Martin-Esteban A, et al. J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 802(2), 347-53, (2004)

Loading...


In vitro metabolism of chlorotriazines: characterization of simazine, atrazine, and propazine metabolism using liver microsomes from rats treated with various cytochrome P450 inducers. Hanioka N, Jinno H, Tanaka-Kagawa T, et al. Toxicol. Appl. Pharmacol. 156(3), 195-205, (1999)

Loading...


Low-density extraction solvent based solvent-terminated dispersive liquid-liquid microextraction for quantitative determination of ionizable pesticides in environmental waters. Tolcha T, Merdassa Y, and Megersa N J. Sep. Sci. 36(6), 1119-27, (2013)

Loading...


Clean-up of triazines in vegetable extracts by molecularly-imprinted solid-phase extraction using a propazine-imprinted polymer. Cacho C, Turiel E, Martín-Esteban A, et al. Anal. Bioanal. Chem 376(4), 491-6, (2003)

Loading...


Detection of pesticides and pesticide metabolites using the cross reactivity of enzyme immunoassays. Thurman EM and Aga DS J. AOAC Int. 84(1), 162-7, (2001)

Loading...


Direct aqueous injection LC-ESI/MS/MS analysis of water for 11 chloro- and thiomethyltriazines and metolachlor and its ethanesulfonic and oxanilic acid degradates. Huang SB, Mayer TJ, and Yokley RA J. Agric. Food Chem. 56(8), 2595-602, (2008)

Loading...


Preservation and analytical procedures for the analysis of chloro-s-triazines and their chlorodegradate products in drinking waters using direct injection liquid chromatography tandem mass spectrometry. Smith GA, Pepich BV, and Munch DJ J. Chromatogr. A 1202(2), 138-44, (2008)

Loading...


Development of a sequential injection chromatography (SIC) method for determination of simazine, atrazine, and propazine. Dos Santos LB, Infante CM, and Masini JC J. Sep. Sci. 32(4), 494-500, (2009)

Loading...


Develop of a multiway chemometric-based analytical method fulfilling regulatory identification criteria: application to GC-MS pesticide residue analysis. Real BD, Ortiz MC, and Sarabia LA J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 910, 122-37, (2012)

Loading...


Remediation of s-triazines contaminated water in a laboratory scale apparatus using zero-valent iron powder. Ghauch A and Suptil J Chemosphere 41(12), 1835-43, (2000)

Loading...


In vitro metabolism of simazine, atrazine and propazine by hepatic cytochrome P450 enzymes of rat, mouse and guinea pig, and oestrogenic activity of chlorotriazines and their main metabolites. Hanioka N, Jinno H, Tanaka-Kagawa T, et al. Xenobiotica 29(12), 1213-26, (1999)

Loading...


Ion-pair association and acid-base equilibria in nonaqueous capillary electrophoresis of weakly basic compounds. Carabias-Martínez R, Rodríguez-Gonzalo E, Hernández-Méndez J, et al. Electrophoresis 27(2), 423-32, (2006)

Loading...


Synthesis and characterization of a propazine imprinted polymer for the extraction of triazines herbicides. Guzzella L, Pozzoni F, and Baggiani C Water Sci. Technol. 57(1), 139-44, (2008)

Loading...


Determination of selected herbicides and phenols in water and soils by solid-phase extraction and high-performance liquid chromatography. Baranowska I and Pieszko C J. Chromatogr. Sci. 38(5), 211-8, (2000)

Loading...


Merck 14,7812

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?