Synonym:
2,4-
CAS Number 1014-70-6
Empirical Formula (Hill Notation) C8H15N5S
Molecular Weight 213.30
Beilstein Registry Number 11728
EC Number 213-801-7
MDL number MFCD00078720
PubChem Substance ID 24869208
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?