CAS Number 5915-41-3
Empirical Formula (Hill Notation) C9H16ClN5
Molecular Weight 229.71
Beilstein Registry Number 3951137
EC Number 227-637-9
MDL number MFCD00055513
PubChem Substance ID 24869235
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?