Synonym: Stirofox
CAS Number 22248-79-9
Empirical Formula (Hill Notation) C10H9Cl4O4P
Molecular Weight 365.96
Beilstein Registry Number 1890909
EC Number 244-865-4
MDL number MFCD00078729
PubChem Substance ID 24869236
| Related Categories | Acaricides, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
| mp | 97-98 °C(lit.) |
| storage temp. | 2-8°C |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?