CAS Number 50471-44-8
Empirical Formula (Hill Notation) C12H9Cl2NO3
Molecular Weight 286.11
Beilstein Registry Number 8331312
EC Number 256-599-6
MDL number MFCD00055511
PubChem Substance ID 24869275
| Related Categories | 2000/60/EC, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | PESTANAL®, analytical standard |
| Gene Information | mouse ... Esr1(13982) |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?