EMAIL THIS PAGE TO A FRIEND
45741 Fluka

Acetic acid

tested according to Ph.Eur.

Synonym: Acidum aceticum glaciale, Glacial acetic acid

Purchase

Properties

Related Categories Acetic Acid, Acids, Acids & Bases, Chemical Synthesis, Essential Chemicals,
vapor density   2.07 (vs air)
autoignition temp.   800 °F
pharmacopeia   testing conforms to Pharmacopeia
expl. lim.   16 %, 92 °F
  4 %, 59 °F
color   colorless
refractive index   n20/D 1.371(lit.)
bp   117-118 °C(lit.)
mp   16.2 °C(lit.)
density   1.049 g/mL at 25 °C(lit.)
Organoleptic   pungent

Description

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH.

General description

Acetic acid is an organic carboxylic acid. It is a pungent smelling hygroscopic liquid. Glacial acetic acid is the anhydrous acetic acid. It forms dimers linked by hydrogen bonds in solution and gaseous commonly referred as phases. It is weakly acidic in nature.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2789PSN1 3(8) / PGII
WGK Germany 
1
RTECS 
AF1225000
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C
Protocols & Articles

Articles

Determination of Water Content in Acetic acid Using Karl Fischer Titration

The determination of water in carboxylic acids is usually straightforward provided the substances are soluble. Strong acids must be neutralized. Acetic acid and related compounds have a tendency to u...
Keywords: Esterifications, Pharmaceutical, Titrations

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?