45935 Fluka

1,2,3-Trimethylbenzene

analytical standard, for environmental analysis

DOWNLOAD MSDS (PDF)

Synonym: Hemellitol

  • CAS Number 526-73-8

  • Linear Formula C6H3(CH3)3

  • Molecular Weight 120.19

  •  Beilstein Registry Number 1903410

  •  EC Number 208-394-8

  •  MDL number MFCD00008520

  •  PubChem Substance ID 24869542

Properties

Related Categories Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard, for environmental analysis
vapor density   4.15 (vs air)
vapor pressure   3.4 mmHg ( 37.7 °C)
autoignition temp.   878 °F
refractive index   n20/D 1.513(lit.)
bp   175-176 °C(lit.)
mp   −25 °C(lit.)
density   0.894 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

1,2,3-<WBR>Trimethylbenzene

technical grade, 90%

1,2,4-<WBR>Trimethylbenzene

analytical standard, for environmental analysis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?