45987 Fluka

(1S)-(+)-Menthyl acetate

analytical standard, for terpene analysis

DOWNLOAD MSDS (PDF)
  • CAS Number 5157-89-1

  • Empirical Formula (Hill Notation) C12H22O2

  • Molecular Weight 198.30

  •  Beilstein Registry Number 1909419

  •  MDL number MFCD00062977

  •  PubChem Substance ID 24869611

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, Chromatography,
grade   analytical standard, for terpene analysis
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]/D +78.5±1°, neat
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.446(lit.)
  n20/D 1.447
bp   228-229 °C(lit.)
density   0.925 g/mL at 25 °C(lit.)

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?