CAS Number 99-09-2
Linear Formula O2NC6H4NH2
Molecular Weight 138.12
Beilstein Registry Number 636962
EC Number 202-729-1
MDL number MFCD00007782
PubChem Substance ID 24869615
| Related Categories | Alpha Sort, Alphabetical Index of Analytical Standards, Amines, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard, for environmental analysis |
| vapor pressure | 1 mmHg ( 119 °C) |
| mp | 111-114 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?