CAS Number 131341-86-1
Empirical Formula (Hill Notation) C12H6F2N2O2
Molecular Weight 248.19
Beilstein Registry Number 8393936
MDL number MFCD01631158
PubChem Substance ID 24869792
Popular Documents: FTNMR (PDF)
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Building Blocks, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?