46208 Fluka

(+)-Fenchone

analytical standard, for terpene analysis

DOWNLOAD MSDS (PDF)

Synonym: (+)-1,3,3-Trimethyl-2-norbornanone, (1S)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one

  • CAS Number 4695-62-9

  • Empirical Formula (Hill Notation) C10H16O

  • Molecular Weight 152.23

  •  Beilstein Registry Number 2206555

  •  EC Number 225-160-0

  •  MDL number MFCD00070689

  •  PubChem Substance ID 24869936

Properties

Related Categories A-F, All Alphabetically Sorted, Alphabetical, Alphabetical Index of Analytical Standards, Analytical Standards,
grade   analytical standard, for terpene analysis
assay   ≥99.5% (sum of enantiomers, GC)
optical activity   [α]20/D +62±1°, neat
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.462
bp   63-65 °C/13 mmHg(lit.)
mp   5-7 °C(lit.)
density   0.945 g/mL at 20 °C(lit.)

Description

Other Notes

Highly purified (+)-fenchone; the keto function can be transformed in many ways1,2

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?