Synonym: (2S)-trans-7-Chloro-2′,4,6-trimethoxy-6′-methylspiro(benzofuran-2[3H],1′-[2]cyclohexene)-3,4′-dione
CAS Number 126-07-8
Empirical Formula (Hill Notation) C17H17ClO6
Molecular Weight 352.77
Beilstein Registry Number 95226
EC Number 204-767-4
MDL number MFCD00082343
PubChem Substance ID 24869893
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Antibiotics, Antibiotics A to Z, |
| grade | VETRANAL™, analytical standard |
| storage temp. | −20°C |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?