Synonym: Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B
CAS Number 548-62-9
Linear Formula C25H30N3Cl
Molecular Weight 407.98
Beilstein Registry Number 3580948
Colour Index Number 42555
EC Number 208-953-6
MDL number MFCD00011750
PubChem Substance ID 24870082
| Related Categories | All alphabetical sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Antibiotics, |
| grade | VETRANAL™, analytical standard |
| mp | 205 °C (dec.)(lit.) |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?