• USA Home
  • 46396 - Malachite Green oxalate salt

EMAIL THIS PAGE TO A FRIEND

46396 Fluka

Malachite Green oxalate salt

VETRANAL, analytical standard

Synonym: N,N,N′,N′-Tetramethyl-4,4′-diaminotriphenylcarbenium oxalate, Basic Green 4

  • CAS Number 2437-29-8

  • Empirical Formula (Hill Notation) C23H25N2 · C2HO4 · 0.5C2H2O4

  • Molecular Weight 463.50

  •  Beilstein Registry Number 3644550

  •  Colour Index Number 42000

  •  EC Number 219-441-7

  •  MDL number MFCD00151209

  •  PubChem Substance ID 24870123

Purchase

Description

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Leucomalachite Green

analytical standard

Malachite Green oxalate salt

Technical grade

Malachite Green oxalate salt

certified by the Biological Stain Commission

Malachite Green oxalate salt

for microscopy, crystalline, S. No.: 754

Malachite Green-d<SUB>5</SUB> picrate

VETRANAL, analytical standard

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36/37-39-46-60-61
RIDADR 
UN 2811 6.1/PG 3
WGK Germany 
3
RTECS 
BQ1190000

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Reversible off-on fluorescence probe for hypoxia and imaging of hypoxia-normoxia cycles in live cells. Takahashi S, Piao W, Matsumura Y, et al. J. Am. Chem. Soc. 134(48), 19588-91, (2012)

Loading...


Biochemical and biophysical response to calcium chloride stress in Aspergillus niger and its role in malachite green degradation. Gomaa OM, Selim NS, and Linz JE Cell Biochem. Biophys. 65(3), 413-23, (2013)

Loading...


Detoxification of malachite green by Pleurotus florida laccase produced under solid-state fermentation using agricultural residues. Sathishkumar P, Palvannan T, Murugesan K, et al. Environ. Technol. 34(1-4), 139-47, (2013)

Loading...


Degradation of malachite green by dielectric barrier discharge plasma. Manoj Kumar Reddy P, Ramaraju B, and Subrahmanyam Ch Water Sci. Technol. 67(5), 1097-104, (2013)

Loading...


High-throughput flow cytometry compatible biosensor based on fluorogen activating protein technology. Wu Y, Tapia PH, Fisher GW, et al. Cytometry. A 83(2), 220-6, (2013)

Loading...


Determination of malachite green, crystal violet and their leuco-metabolites in fish by HPLC-VIS detection after immunoaffinity column clean-up. Xie J, Peng T, Chen DD, et al. J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 913-914, 123-8, (2013)

Loading...


Hazardous dyes removal from aqueous solution over mesoporous aluminophosphate with textural porosity by adsorption. Kannan C, Muthuraja K, and Devi MR J. Hazard. Mater. 244-245, 10-20, (2013)

Loading...


Preparation of acrylic acid-modified chitin improved by an experimental design and its application in absorbing toxic organic compounds. Huang CM, Chen LC, Yang HC, et al. J. Hazard. Mater. 241-242, 190-6, (2012)

Loading...


Analyses of enrofloxacin, furazolidone and malachite green in fish products with surface-enhanced Raman spectroscopy. Zhang Y, Huang Y, Zhai F, et al. Food Chem. 135(2), 845-50, (2012)

Loading...


Ultrasonic-assisted ozone oxidation process of triphenylmethane dye degradation: evidence for the promotion effects of ultrasonic on malachite green decolorization and degradation mechanism. Zhou XJ, Guo WQ, Yang SS, et al. Bioresour. Technol. 128, 827-30, (2013)

Loading...


Application of ionic liquid-based microwave-assisted extraction of malachite green and crystal violet from water samples. Gao Z, Liu T, Yan X, et al. J. Sep. Sci. 36(6), 1112-8, (2013)

Loading...


Application of graphene-based solid-phase extraction for ultra-fast determination of malachite green and its metabolite in fish tissues. Chen L, Lu Y, Li S, et al. Food Chem. 141(2), 1383-9, (2013)

Loading...


Time-resolved molecular transport across living cell membranes. Zeng J, Eckenrode HM, Dounce SM, et al. Biophys. J. 104(1), 139-45, (2013)

Loading...


Malachite green mediates homodimerization of antibody VL domains to form a fluorescent ternary complex with singular symmetric interfaces. Szent-Gyorgyi C, Stanfield RL, Andreko S, et al. J. Mol. Biol. 425(22), 4595-613, (2013)

Loading...


A novel polymerization of ultrathin sensitive imprinted film on surface plasmon resonance sensor. Dong J, Peng Y, Gao N, et al. Analyst 137(19), 4571-6, (2012)

Loading...


A malachite green-based assay to assess glucan phosphatase activity. Sherwood AR, Paasch BC, Worby CA, et al. Anal. Biochem. 435(1), 54-6, (2013)

Loading...


Lighting-up of the dye malachite green with mercury(II)-DNA and its application for fluorescence turn-off detection of cysteine and glutathione. Jia X, Li J, and Wang E Chemistry 18(42), 13494-500, (2012)

Loading...


Catalytic DNA with phosphatase activity. Chandrasekar J and Silverman SK Proc. Natl. Acad. Sci. U. S. A. 110(14), 5315-20, (2013)

Loading...


Molecularly imprinted polymer for selective extraction of malachite green from seawater and seafood coupled with high-performance liquid chromatographic determination. Lian Z and Wang J Mar. Pollut. Bull. 64(12), 2656-62, (2012)

Loading...


Malachite green derivative-functionalized single nanochannel: light-and-pH dual-driven ionic gating. Wen L, Liu Q, Ma J, et al. Adv. Mater. 24(46), 6193-8, (2012)

Loading...


Vfa1 binds to the N-terminal microtubule-interacting and trafficking (MIT) domain of Vps4 and stimulates its ATPase activity. Vild CJ and Xu Z J. Biol. Chem. 289(15), 10378-86, (2014)

Loading...


Thermodynamics and kinetics of adaptive binding in the malachite green RNA aptamer. Da Costa JB, Andreiev AI, and Dieckmann T Biochemistry 52(38), 6575-83, (2013)

Loading...


Streptavidin binding bifunctional aptamers and their interaction with low molecular weight ligands. Le TT, Scott S, and Cass AE Anal. Chim. Acta 761, 143-8, (2013)

Loading...


Pathway and molecular mechanisms for malachite green biodegradation in Exiguobacterium sp. MG2. Wang J, Gao F, Liu Z, et al. PLoS ONE 7(12), e51808, (2012)

Loading...


Fluorogenic RNA nanoparticles for monitoring RNA folding and degradation in real time in living cells. Reif R, Haque F, and Guo P Nucleic Acid Ther. 22(6), 428-37, (2012)

Loading...


Preparation of char from lotus seed biomass and the exploration of its dye removal capacity through batch and column adsorption studies. Nethaji S, Sivasamy A, Kumar RV, et al. Environ. Sci. Pollut. Res. Int. 20(6), 3670-8, (2013)

Loading...


A nanosilver-based spectrophotometric method for determination of malachite green in surface water samples. Sahraei R, Farmany A, Mortazavi SS, et al. Environ. Monit. Assess. 185(7), 5817-22, (2013)

Loading...


Determination of residues of three triphenylmethane dyes and their metabolites (malachite green, leuco malachite green, crystal violet, leuco crystal violet, and brilliant green) in aquaculture products by LC/MS/MS: first action 2012.25. Hurtaud-Pessel D, Couëdor P, Verdon E, et al. J. AOAC Int. 96(5), 1152-7, (2013)

Loading...


Adsorption of neutral red and malachite green onto grapefruit peel in single and binary systems. Zou W, Gao S, Zou X, et al. Water Environ. Res. 85(5), 466-77, (2013)

Loading...


Purification and characterization of a novel laccase from Coprinus cinereus and decolorization of different chemically dyes. Lin Y, Zhang Z, Tian Y, et al. Mol. Biol. Rep. 40(2), 1487-94, (2013)

Loading...


Merck 14,5699

Beil. 13,IV,2279

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?