CAS Number 74223-64-6
Empirical Formula (Hill Notation) C14H15N5O6S
Molecular Weight 381.36
Beilstein Registry Number 587472
MDL number MFCD00128061
PubChem Substance ID 24870165
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | PESTANAL®, analytical standard |
PESTANAL is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?