Synonym:
1,3,5(10)
CAS Number 50-27-1
Empirical Formula (Hill Notation) C18H24O3
Molecular Weight 288.38
Beilstein Registry Number 2508172
EC Number 200-022-2
MDL number MFCD00003691
PubChem Substance ID 24870296
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Bioactive Small Molecules, |
| grade | VETRANAL™, analytical standard |
| mp | 280-282 °C(lit.) |
| Gene Information |
human ... SERPINA6(866) mouse ... Esr1(13982) rat ... Ar(24208) |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?