CAS Number 2058-46-0
Empirical Formula (Hill Notation) C22H24N2O9 · HCl
Molecular Weight 496.89
Beilstein Registry Number 3853107
E Number E703
EC Number 218-161-2
MDL number MFCD00135815
PubChem Substance ID 24870318
| Related Categories | 1694 Pharmaceuticals & Personal Care Products, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Antibacterial, |
| grade | VETRANAL™, analytical standard |
| suitability | suitable for 1694 per US EPA |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?