CAS Number 22189-32-8
Empirical Formula (Hill Notation) C14H24N2O7 · 2HCl · 5H2O
Molecular Weight 495.35
Beilstein Registry Number 5684150
EC Number 244-554-3
MDL number MFCD00150886
| Related Categories | Alphabetical Index of Analytical Standards, Aminoglycosides, Analytical Standards, Analytical/Chromatography, Antibacterial, |
| grade | VETRANAL™, analytical standard |
| storage temp. | 2-8°C |
VETRANAL is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Ready Made Solution, 100 mg/mL in DMSO/H2O, 1:1, 0.2 μm filtered
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?