46984 Fluka

Trimethoprim

VETRANAL, analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine

  • CAS Number 738-70-5

  • Empirical Formula (Hill Notation) C14H18N4O3

  • Molecular Weight 290.32

  •  Beilstein Registry Number 625127

  •  EC Number 212-006-2

  •  MDL number MFCD00036761

  •  PubChem Substance ID 24870616

Description

Application

Commission Regulation (EU) No 37/2010 of 22 December 2009 on pharmacologically active substances and their classification regarding maximum residue limits in foodstuffs of animal origin

Primarily used as an antibacterial agent. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme.

Other Notes

Specific inhibitor of formulation. Widely used in studies on dihydrofolate reductase1,2.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Sulfamethoxazole

VETRANAL, analytical standard

Trimethoprim

≥98% (TLC)

Trimethoprim

crystallized, ≥99.0% (HPLC)

Trimethoprim

pharmaceutical secondary standard; traceable to USP, PhEur and BP

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?