Synonym: HF
CAS Number 7664-39-3
Empirical Formula (Hill Notation) HF
Molecular Weight 20.01
MDL number MFCD00011346
PubChem Substance ID 24871149
| Related Categories | Acids & Bases, Alphabetical Index of Analytical Standards, Analytical Reagents, Analytical Standards, Analytical/Chromatography, |
| grade | for trace analysis |
| vapor density | 1.27 (vs air) |
| vapor pressure | 25 mmHg ( 20 °C) |
| grade | TraceSELECT® |
| assay | 47-51% (acidimetric) |
| 47-51% | |
| density | 1.15 g/mL at 25 °C(lit.) |
100, 500 mL in poly bottle
TraceSELECT is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?