487139 Fluka

Gadolinium(III) chloride solution

NMR reference standard, water 1 %, DSS 0.1 %, 0.1 mg/mL in D2O (99.9 atom % D)

DOWNLOAD MSDS (PDF)
  • Linear Formula GdCl3

  • Molecular Weight 263.61

  •  MDL number MFCD00011024

Properties

Related Categories Alphabetical Index of Analytical Standards, Alphabetical Listings, Analytical Standards, Analytical/Chromatography, Catalysis and Inorganic Chemistry,
grade   NMR reference standard
composition   DSS, 0.1%
  water, 1%
concentration   0.1 mg/mL in D2O (99.9 atom % D)
NMR tube O.D. × L   5 mm × 8 in.

Description

Features and Benefits

Auto test

Price and Availability

Request a Quote for this
Stable Isotope Product


Isotec Stable IsotopesRequest Stable Isotope product pricing
and availablity.






Customers Also Viewed

Gadolinium(III) chloride

anhydrous, powder, 99.99% trace metals basis

Gadolinium(III) chloride hexahydrate

99.999% trace metals basis

Gadolinium(III) chloride solution

NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), DSS 0.1 %, water 1 %

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?