MDL number MFCD00000826
PubChem Substance ID 24872248
| Related Categories | Alphabetical Index of Analytical Standards, Alphabetical Listings, Analytical Standards, Analytical/Chromatography, C, |
| grade | NMR reference standard |
| concentration | 1% in acetone-d6 (99.9 atom % D) |
| NMR tube O.D. | 5 mm |
1H line shape
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?